3-Methyl-2-butenyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic Acid Prenyl EsterPrenyl Acetate3-Methylbut-2-en-1-yl acetateAcetic Acid 3-Methyl-2-butenyl Ester

A1148-500ML A1148-25ML
TCA1148-500MLEA 199 USD
TCA1148-500ML TCA1148-025ML
3-Methyl-2-butenyl acetate ≥98.0%
3-Methyl-2-butenyl acetate

CAS Number: 1191-16-8
MDL Number: MFCD00036569
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Acetic Acid 3-Methyl-2-butenyl Ester, Acetic Acid Prenyl Ester, Prenyl Acetate

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 128 °C
Density: 0.92 (20 °C)
MDL Number: MFCD00036569
CAS Number: 1191-16-8
https://us.vwr.com/stibo/web/std.lang.all/89/93/28928993.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR