Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Acetyl-2,5-dimethylfuran ≥98.0%

Supplier: TCI America


A1121-25G
TCA1121-025GEA 58.3 USD InStock
TCA1121-025G
3-Acetyl-2,5-dimethylfuran ≥98.0%
3-Acetyl-2,5-dimethylfuran
CAS Number: 10599-70-9
MDL Number: MFCD00010281
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 82
Specific Gravity (20/20): 1.04


Formula: C₈H₁₀O₂
MW: 138,17 g/mol
Boiling Pt: 196 °C
Density: 1,04 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00010281
CAS Number: 10599-70-9
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty