(R)-(-)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
Synonyms:
(R)-Pyrrolidin-3-amine dihydrochloride
A1053-5G
A1053-25G
TCA1053-025GEA
989.83
USD
TCA1053-025G
TCA1053-005G
(R)-(-)-3-Aminopyrrolidine dihydrochloride ≥98.0% (by titrimetric analysis)
(R)-(-)-3-Aminopyrrolidine dihydrochloride
CAS Number: 116183-81-4
MDL Number: MFCD00070604
Molecular Formula: C4H10N2
Molecular Weight: 159.05
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -1.1 deg (C=10, H2O)
Formula:
C₄H₁₂Cl₂N₂ MW: 159.06 g/mol |
MDL Number:
MFCD00070604 CAS Number: 116183-81-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...