Aminoacetonitrile sulfate ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: Cyanomethylamine SulfateCyanomethylamine Sulphate

A0912-500G A0912-25G
TCA0912-500GEA 422.61 USD
TCA0912-500G TCA0912-25G
Aminoacetonitrile sulfate ≥98.0% (by total nitrogen and titration analysis)
Aminoacetonitrile sulfate

CAS Number: 5466-22-8
MDL Number: MFCD00012851
Molecular Formula: C2H4N2
Molecular Weight: 210.21
Purity/Analysis Method: 98.0% (N,T)
Form: Crystal
Melting point (°C): 165


Other Product Names / Synonyms: Cyanomethylamine Sulfate

Formula: C₂H₄N₂
Melting Pt: 165 °C
MDL Number: MFCD00012851
CAS Number: 5466-22-8
https://us.vwr.com/stibo/web/std.lang.all/86/60/28928660.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR