Acetophenone oxime ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

Synonyms: 1-Phenyl-1-ethanone oximeMethyl phenyl ketoxime

A0807-25G
TCA0807-25GEA 60 USD
TCA0807-25G
Acetophenone oxime ≥98.0% (by GC, total nitrogen)
Acetophenone oxime

Form: Crystal

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 119 °C
Melting Pt: 60 °C
MDL Number: MFCD00013931
CAS Number: 613-91-2
EINECS: 210-360-2
https://us.vwr.com/stibo/web/std.lang.all/85/51/28928551.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR