Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Abscisic Acid (Synthetic) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America


A0792-500MG A0792-100MG
TCA0792-500MGEA 274.75 USD InStock
TCA0792-500MG TCA0792-100MG
Abscisic Acid (Synthetic) ≥98.0% (by HPLC, titration analysis)
Abscisic acid (synthetic)
CAS Number: 14375-45-2
MDL Number: MFCD00075619
Molecular Formula: C15H20O4
Molecular Weight: 264.32
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 186
Storage Temperature: 0-10°C


Formula: C₁₅H₂₀O₄
MW: 264.32 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00075619
CAS Number: 14375-45-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty