1-Acetamidoadamantane ≥98.0%

Supplier: TCI America

Synonyms: N-Acetyl-1-adamantanamineN-1-Adamantylacetamide

A0737-25G
TCA0737-025GEA 76 USD
TCA0737-025G
1-Acetamidoadamantane ≥98.0%
1-Acetamidoadamantane

CAS Number: 880-52-4
MDL Number: MFCD00074730
Molecular Formula: C12H19NO
Molecular Weight: 193.29
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 148


Other Product Names / Synonyms: N-Acetyl-1-adamantanamine, N-1-Adamantylacetamide

Formula: C₁₂H₁₉NO
MW: 193.29 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00074730
CAS Number: 880-52-4
https://us.vwr.com/stibo/web/std.lang.all/84/18/28928418.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR