Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-D-leucine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ac-D-Leu-OH

A0713-1G
TCA0713-001GEA 138.37 USD InStock
TCA0713-001G
N-Acetyl-D-leucine ≥99.0% (by titrimetric analysis)
N-Acetyl-D-leucine
CAS Number: 19764-30-8
MDL Number: MFCD00066069
Molecular Formula: C8H15NO3
Molecular Weight: 173.21
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 23 deg (C=4, EtOH)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Ac-D-Leu-OH


Formula: C₈H₁₅NO₃
MW: 173,2 g/mol
MDL Number: MFCD00066069
CAS Number: 19764-30-8
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty