Azo rubine
Supplier: TCI America
Synonyms:
acid Red 14
, Brilliant Crimson Red
, Carmoisine B
, Sodium 4-hydroxy-3-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate
, Sodium 4-hydroxy-3-((4-sulfonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulfonate
A0580-25G
TCA0580-025GEA
51.74
USD
TCA0580-025G
Azo rubine
Azo rubine
CAS Number: 3567-69-9
MDL Number: MFCD00003978
Molecular Formula: C20H14N2O7S2
Molecular Weight: 502.42
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: Brilliant Crimson Red, Carmoisine B
Formula:
C₂₀H₁₄N₂O₇S₂ MW: 502.42 g/mol |
MDL Number:
MFCD00003978 CAS Number: 3567-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...