6-Azathymine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A0556-1G
TCA0556-001GEA 70 USD
TCA0556-001G
6-Azathymine ≥98.0% (by HPLC, titration analysis)
6-Azathymine

CAS Number: 932-53-6
MDL Number: MFCD00006457
Molecular Formula: C4H5N3O2
Molecular Weight: 127.10
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214

Formula: C₄H₅N₃O₂
Melting Pt: 214 °C
MDL Number: MFCD00006457
CAS Number: 932-53-6
https://us.vwr.com/stibo/web/std.lang.all/79/86/28927986.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR