Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxydiphenylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America


A0476-25G
TCA0476-025GEA 60.56 USD InStock
TCA0476-025G
3-Hydroxydiphenylamine ≥98.0% (by HPLC, total nitrogen)
3-Hydroxydiphenylamine
CAS Number: 101-18-8
MDL Number: MFCD00002262
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Boiling point (°C): 340
Melting point (°C): 80

Other Product Names / Synonyms: m-Anilinophenol, N-(3-Hydroxyphenyl)aniline


Formula: C₁₂H₁₁NO
MW: 185,23
Boiling Pt: 340 °C
Melting Pt: 80 °C
MDL Number: MFCD00002262
CAS Number: 101-18-8
EINECS: 202-923-6
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,N)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty