1-Amino-4-naphthol hydrochloride ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Amino-1-naphthol hydrochloride
A0366-5G
TCA0366-5GEA
140.07
USD
TCA0366-5G
1-Amino-4-naphthol hydrochloride ≥98.0% (by HPLC)
1-Amino-4-naphthol hydrochloride
CAS Number: 5959-56-8
MDL Number: MFCD00012562
Molecular Formula: C10H9NO
Molecular Weight: 195.65
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 280
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...