2-Amino-4-chloro-5-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
A0286-25G
TCA0286-025GEA
31.45
USD
TCA0286-025G
2-Amino-4-chloro-5-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-4-chloro-5-methylbenzenesulphonic acid
CAS Number: 88-51-7
MDL Number: MFCD00035771
Molecular Formula: C7H8ClNO3S
Molecular Weight: 221.66
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 5-Chloro-4-methylaniline-2-sulfonic Acid, 5-Chloro-p-toluidine-2-sulfonic Acid, 2-Amino-4-chloro-5-methylbenzenesulfonic Acid
Formula:
C₇H₈ClNO₃S MW: 221.6613 g/mol |
MDL Number:
MFCD00035771 CAS Number: 88-51-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...