Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropyl 4-aminobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America


A0273-25G
TCA0273-025GEA 52.03 USD InStock
TCA0273-025G
Isopropyl 4-aminobenzoate ≥98.0% (by GC, titration analysis)
Isopropyl 4-aminobenzoate
CAS Number: 18144-43-9
MDL Number: MFCD00017108
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 84

Other Product Names / Synonyms: 4-Aminobenzoic Acid Isopropyl Ester


Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty