Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Acetylurea ≥99.0% (by total nitrogen basis)

Supplier: TCI America


A0124-25G
TCA0124-025GEA 78.03 USD InStock
TCA0124-025G
Acetylurea ≥99.0% (by total nitrogen basis)
Acetylurea
CAS Number: 591-07-1
MDL Number: MFCD00038140
Molecular Formula: C3H6N2O2
Molecular Weight: 102.09
Purity/Analysis Method: 99.0% (N)
Form: Crystal
Melting point (°C): 215


Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00038140
CAS Number: 591-07-1
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (N)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty