NXP 150 Z-Flex Aluminized Heat Resistant Sleeves, Newtex
Supplier: Newtex
2100106
75794-926CS 908.97 USD
75794-926
NXP 150 Z-Flex Aluminized Heat Resistant Sleeves, Newtex
Protection from short duration ambient heat up to 93 °C (200 °F) and radiant heat up to 1093 °C (2000 °F).
  • Made from Z-Flex multilayer aluminized technology, which reflects 95% of radiant heat
  • Commonly used in conjunction with heat resistant gloves to extend protection up the arm
Common applications: Foundries and casting operations, welding and hot works operations, glass and ceramic plants production, power generation, petrochemical and refining, laboratory and testing facilities. 38.1 cm (15") length. Made in the USA.
Certifications: Z-Flex materials have been tested in accordance with ASTM F955, ASTM F1939, EN 532, EN 367, EN 366, EN 407, and MIL-C-24929.