Upchurch Scientific® PEEK™ Spiral-Link™ Tubing, IDEX Health & Science
17110 17101 17304 17305 17402 17405 17105 17301 17210 17302 17204 17205 17320 17104 17410 17220 17420 17120 17404 17310 17102 17401
21001-140EA 28.83 USD
21001-140 53498-584 21001-152 21001-130 53498-586 21001-150 53498-588 21001-144 21001-134 21511-604 21001-142 21001-132 21001-154 53498-578 21001-126 21001-148 21001-138 21001-146 53498-582 21001-128 53498-592 21511-420
Upchurch Scientific® PEEK™ Spiral-Link™ Tubing, IDEX Health & Science
Upchurch Scientific® PEEK™ polymer Spiral-Link™ tubing is preformed into convenient coils, allowing easy movement of system components and equipment repairs whenever needed without the hassle of breaking connections
Upchurch Scientific® PEEK™ (polyetheretherketone) polymer tubing is biocompatible, chemically inert to most solvents, and can be used to replace stainless steel tubing in most liquid analytical systems. Unlike stainless steel tubing, PEEK™ tubing is flexible and can be easily cut to desired lengths. PEEK™ tubing can be used with stainless steel or polymer fittings. PEEK™ tubing has a very smooth internal surface, which causes less turbulence than similarly sized metal tubing, contributing to improved resolution of sample bands. Of all the Upchurch Scientific® polymer tubing materials, PEEK™ is the least permeable to gas. Please see the Specifications tab for more information.
These 1/16" O.D. tubing products can also be used as sample loops. Spiral-Links™ come in multiple volumes and lengths, and they are pressure rated to 345 bar (5,000 psi).
Certifications: These products are RoHS compliant.
Ordering information: Each Spiral-Link™ ships with two Upchurch Scientific® F-287 SealTight Fittings (VWR # 21511-861).