Anti-ZAP70 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12869
89359-526EA 582.56 USD
89359-526
Anti-ZAP70 Rabbit Polyclonal Antibody
Antibodies
ZAP70, a 70 kDa member of the Syk tyrosine kinase family, plays a central role in lymphocyte activation and development, and is implicated in several immune disorders. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling. Phosphorylation of tyrosines 315 and 319 plays an important role in mediating the positive and negative selection of T cells in thymus.

WB: Use at a dilution of 1/1000. Predicted molecular weight: 74.3 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ZAP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR