Anti-Y14 Mouse Monoclonal Antibody [clone: 4C4]

Supplier: GeneTex
GTX25828
89361-754EA 611.49 USD
89361-754
Anti-Y14 Mouse Monoclonal Antibody [clone: 4C4]
Antibodies
Mouse monoclonal [4C4] to Y14

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 21 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Y14
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR