Anti-XRCC3 Mouse Polyclonal Antibody

Supplier: GeneTex
GTX70292
89364-860EA 579.79 USD
89364-860
Anti-XRCC3 Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antiserum to human XRCC3

For WB: Use at a dilution of 1:500-1,000. Block with TBST/3% non-fat milk. Optimal working concentration for specific experiments needs to be determined by researcher.

Type: Primary
Antigen: XRCC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR