Anti-XRCC1 Mouse Monoclonal Antibody [clone: 33-2-5]

GTX23133
89351-888EA 376.91 USD
89351-888
Anti-XRCC1 Mouse Monoclonal Antibody [clone: 33-2-5]
Mouse monoclonal [33-2-5] to XRCC 1

For IP (Native, with protein A): Use at a dilution of 1:400 protein lysate. For WB: Use at a dilution of 1:200. For IHC-P: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: XRCC1
Clonality: Monoclonal
Clone: 33-2-5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR