Anti-XAB2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70313
89364-896EA 579.79 USD
89364-896
Anti-XAB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to human XAB2 ( carboxy terminus )

Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: XAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR