Anti-WNT5A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100618
89348-570EA 631.13 USD
89348-570
Anti-WNT5A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to WNT5A (wingless-type MMTV integration site family, member 5A)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: WNT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR