Anti-VSV Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23556
89352-026EA 601.14 USD
89352-026
Anti-VSV Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VSV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR