Anti-VIM Mouse Monoclonal Antibody

Supplier: Genetex

GTX27752
89352-288EA 395.31 USD
89352-288
Anti-VIM Mouse Monoclonal Antibody
Antibodies
Vimentin is the major subunit protein of the intermediate filaments of mesenchumal cells. Immunohistochemical staining for Vimentin is thus characteristic of sarcomas (of neural, muscle and fibroblast origin) compared to carcinomas which are generally negative. Melanomas, lymphomas and vascular tumors may all stain for Vimentin. Vimentin antibodies are thus of value in the differential diagnosis of malignant tumors, generally used with a panel of other antibodies including those recognising cytokeratins, lymphoid markers, S100, desmin and neurofilaments.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR