Anti-VDAC2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX104745
89350-516EA 404.77 USD
89350-516
Anti-VDAC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to VDAC2 (voltage-dependent anion channel 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: VDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR