Anti-URM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24755
89367-580EA 862.99 USD
89367-580
Anti-URM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Urm1

Recommended Starting Dilutions:WB: Use at 1:500 - 1:2,000ELISA: Use at 1:2,000 - 1:10,000

Type: Primary
Antigen: URM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: S. cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR