Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99]

Supplier: GeneTex
GTX23312
89367-070EA 726 USD
89367-070
Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99]
Antibodies
Mouse monoclonal [TA99] to Tyrosinase Related Protein 75

Not tested in applications other than those listed above. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of 75 kDa, which corresponds to the predicted molecular weight of Tyrosinase Related Protein 75.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR