Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tubulin Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11270
The major building block of microtubules is tubulin, an intracellular cylindrical filamentous structure that is present in almost all eukaryotic cells. Except in the simplest eukaryotes, tubulin (100 kDa) exists in all cells as a heterodimer of two similar but non-identical polypeptides (55 kDa each), designated alpha and beta, that assemble into microtubules. Within either family of alpha/beta tubulin heterodimers, individual subunits diverge from each other (both within and across species) at less than 10% of the amino acid positions. The most extreme diversity is localized to the carboxyl-terminal 15 residues. Both alpha and beta tubulins consist of various isotypes. In addition, both undergo post-translational modifications, including acetylation, phosphorylation detyrosination, polyglutamylation, and polyglycylation. Polyglutamylation of tubulin consists of the addition of a lateral chain of glutamyl units on the C-terminal region of tubulin polypeptides. This modification was shown to regulate the interaction between tau, one of the major neuronal microtubule associated proteins, and tubulin. Furthermore, it was shown that injection of monoclonal antibodies specific to the polyglutamylated form of tubulin into HeLa cells caused the disappearance of centrioles, an organelle known to have the most stable microtubules. The detection, localization, and characterization of proteins involved in microtubule function is fundamental to the understanding of mitosis, meiosis, organellar and flagellar movement, intracellular transport, and cytoskeletal functions. Antibodies reacting specifically with the modified forms of alpha and beta tubulin isotypes serve as essential tools in the detection and study of the functional significance of these molecules.

Type: Primary
Antigen: Tubulin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:




 

ORDER

Expand All / Collapse All
Supplier No. VWR Catalog Number Unit Price Quantity
GTX11270 89357-156 Each (200µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TUB
Antigen Name Tubulin
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Immunogen Protein purified from 13 day old chicken embryos.
Purification Whole antiserum
Storage Buffer Preservative: 0.1% Sodium Azide
Storage Temperature Aliquot and store at -20°C or -80°C. Avoid repeated freeze / thaw cycles.
Tested Applications ICC/IF, WB

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty