Anti-TTF1 Mouse Monoclonal Antibody [clone: 8G7G3/1]

Supplier: GeneTex
GTX73528
89353-198EA 617.59 USD
89353-198
Anti-TTF1 Mouse Monoclonal Antibody [clone: 8G7G3/1]
Antibodies
Mouse monoclonal antibody [8G7G3/1] to NKX2.1

This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.

Type: Primary
Antigen: TTF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR