Anti-TRPV1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX80788
89366-690EA 588.23 USD
89366-690
Anti-TRPV1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TRPV1 antiserum

Immunohistochemistry 1:1000 Dilutions listed only as a recommendation. Optimal dilution should be determined by investigator. Serum was screened on mouse DRG by immunohistochemistry. Tissue was fixed with 2% paraformaldehyde.

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR