Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRPC4 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX12247
89359-298EA 380.82 USD InStock
89359-298
Anti-TRPC4 Rabbit Polyclonal Antibody
Antibodies
The TRP isoforms can be divided into three subfamilies based on characteristic sequence domains.The long TRP channels (LTRPCs) have four related sequences with open reading frames (ORFs) coding for approximately 1600 amino acids. The other two subfamilies are characterized by shorter ORFs, coding for about 900 amino acids. These are called the short TRP(STRP) and osm-9-like (OTRP) channel families. TRPC4 has been identified in adrenal gland, testis and brain. Full-length cDNAs have been cloned from bovine, human, mouse and rat species. TRPC4 is a non-selective cation channel that integrates signaling pathways from G-protein-coupled receptors and receptor tyrosine kinases independently of the depletion of calcium stores.

WB: Use at a dilution of 1/200. Predicted molecular weight: 117 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TRPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty