Anti-TRIB3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX42107
89363-494EA 602.26 USD
89363-494
Anti-TRIB3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TRIB3

WB: Use at a concentration of 1 - 3 ?g/ml. Detects a band of approximately 40 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TRIB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR