Anti-Treponema Pallidum Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX40465
89363-194EA 481.95 USD
89363-194
Anti-Treponema Pallidum Rabbit Polyclonal Antibody
Antibodies
Treponema pallidum is the causative agent of syphilis. It is a spirochete, a helical to sinusoidal bacterium with outer and cytoplasmic membranes, a thin peptidoglycan layer, and periplasmic flagella.

Type: Primary
Antigen: Treponema Pallidum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR