Anti-TF Mouse Monoclonal Antibody [clone: 11D3]

Supplier: GeneTex
GTX10208
89350-694EA 759 USD
89350-694
Anti-TF Mouse Monoclonal Antibody [clone: 11D3]
Antibodies
Mouse monoclonal [11D3] to Transferrin

ELISA: Use at an assay dependent dilution. Can be used in ELISA for the detection of tranferrin in serum by sandwich ELISA as an antibody pair with GTX10212 as the capture antibody and using GTX10208 for detection. Can be used for the detection of transferrin-transferrin receptor complex by ELISA in combination with an anti-transferrin recptor antibody. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone: 11D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR