Anti-TF Chicken Polyclonal Antibody (HRP)

GTX77516
89353-762EA 235.16 USD
89353-762
Anti-TF Chicken Polyclonal Antibody (HRP)
HRP Conjugated Chicken anti - Human Transferrin

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR