Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF-related Apoptosis-inducing Ligand Mouse Monoclonal Antibody [clone: 200000]

Supplier: Genetex
GTX22219
Apoptosis or programmed cell death is induced in cells by a group of death domain-containing receptors. Binding of ligand to these receptors sends signals that activate members of the caspase family of proteases. The signals ultimately cause degradation of chromosomal DNA by activating DNase. TRAIL (TNF-related apoptosis-induced ligand) or Apo-2L initiates apoptosis of tumor cells by binding to either of its receptors, DR4 or DR5. These receptors consist of an extracellular TRAIL binding domain and a cytoplasmic "death domain". In addition, two decoy receptors for TRAIL have also been identified. These receptors, designated DcR1 and DcR2, lack the death domain. Binding of TRAIL to either of these receptors, therefore, does not transmit the death signal. Thus, these receptors represent a novel way of regulating cell sensitivity to a pro-apoptotic cytokine at the cell surface. TRAIL is expressed predominantly in spleen, lung, and prostate but also in many other tissues. TRAIL (also called Apo-2 ligand) is a type II membrane protein from the TNF family. The human form of the protein is 281 amino acids in length, whereas the murine form is 291 amino acids. The two forms share 65% identity. Recombinant human TRAIL monomer has a molecular mass of approximately 21 kDa. Due to glycosylation, recombinant human TRAIL migrates as a 24 kDa protein in SDS-PAGE.

FuncS: Use at an assay dependent concentration. Has high neutralizing activity for human TRAIL in biological assays. Neutralizing activity was tested in Jurkat T cells, where apoptosis was induced by a recombinant human soluble TRAIL (300 ng/ml), it completely neutralizes rhsTRAIL mediated killing in a concentration range of 0.5-1.5μg/ml). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Monoclonal
Clone: 200000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg GTX22219 89361-192 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TNFSF10
Antigen Name Tumor Necrosis Factor (Ligand) Superfamily, Member 10
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 2E5
Host Mouse
Isotype IgG1
Antigen Synonyms TNF-related apoptosis-inducing ligand, Tumor necrosis factor ligand superfamily member 10, Apo-2L, Protein TRAIL, CD253, Apo-2 ligand
Amino Acid Number 95 to 281
Gene ID 8743
Concentration 1 mg/ml
Immunogen Recombinant soluble fragment, corresponding to amino acids 95-281 of Human TRAIL.
Purification >95% by SDS-PAGE
Storage Buffer Preservative: NoneConstituents: PBS. pH 7.4
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty