Anti-TRAF3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59903
89363-752EA 528.9 USD
89363-752
Anti-TRAF3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TRAF3

Western blotting (0.5-4 µg/ml). However, the optimal conditions should be determined individually. The antibody recognizes 67 kDa TRAF3 of human, mouse and rat origins. Reactivity to other species has not been tested.

Type: Primary
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR