Anti-TPA Sheep Polyclonal Antibody

Supplier: GeneTex
GTX79606
89366-646EA 611.84 USD
89366-646
Anti-TPA Sheep Polyclonal Antibody
Antibodies
Sheep Polyclonal antibody to tPA (HRP)

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR