Anti-TNFRSF1B Goat Polyclonal Antibody

Supplier: GeneTex
GTX10502
89354-066EA 660.25 USD
89354-066
Anti-TNFRSF1B Goat Polyclonal Antibody
Antibodies
Goat polyclonal to TNFRSF1B

ELISA: Use at a concentration of 0.5 - 1 ?g/ml, this is determined to detect recombinant human sTNF RII to a limit of 78 pg/well. WB: Use at a concentration of 1 - 2 ?g/ml, this is determined using recombinant human sTNF RII at 5 ng/lane under non-reducing conditions. Predicted molecular weight: 75 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNFRSF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR