Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF14 Rat Monoclonal Antibody

Supplier: Genetex

GTX16093
89360-158EA 380.79 USD InStock
89360-158
Anti-TNFRSF14 Rat Monoclonal Antibody
Antibodies
TNFRSF14, also known as HVEM (herpes virus entry mediator), TR2 (TNF receptor like molecule) and ATAR (another TRAF-associated receptor), is a type I membrane protein belonging to the TNF receptor superfamily. This receptor mediates herpes virus entry into cells during infection. TNFRSF14 is able to inhibit the proliferation, activation, and cytokine production of T cells. It has an extracellular domain containing several cysteine-rich repeats and a short cytoplasmic region containing a TRAF (TNF receptor-associated factor) interaction domain. The extracellular domain of TNFRSF14 interacts with the herpes simplex virus envelope glycoprotein D. TNFRSF14 binds two cellular ligands: lymphotoxin alpha and LIGHT. LIGHT is an acronym, which stands for is homologous to lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes. The LIGHT:TNFRSF14 interaction controls immune response functions by cell death induction as well as cell activation. TNFRSF14 is expressed by peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells.

FACS: Use at a concentration of 1 μg/ml. IF: Use at a dilution of 1/1000. IP: Use at a dilution of 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNFRSF14
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty