Anti-TNFA Mouse Monoclonal Antibody [clone: TA20]

Supplier: GeneTex
GTX10205
89350-692EA 726.31 USD
89350-692
Anti-TNFA Mouse Monoclonal Antibody [clone: TA20]
Antibodies
Mouse monoclonal [TA20] to TNF alpha

FuncS: Use at an assay dependent dilution. Inhibits the biological activity of TNF alpha in different biological assays. WB: Use at an assay dependent dilution. TNF alpha is normally secreted as a homotrimer with a molecular mass of 52 kDa. Monomeric TNF-alpha (17.4 kDa) is not biologically active Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: TA20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR