Anti-TM4SF4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX46435
89363-610EA 585.66 USD
89363-610
Anti-TM4SF4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TM4SF4 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TM4SF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR