Anti-TLR2 Mouse Monoclonal Antibody [clone: 11G7]

Supplier: GeneTex
GTX42127
89363-496EA 542.97 USD
89363-496
Anti-TLR2 Mouse Monoclonal Antibody [clone: 11G7]
Antibodies
Mouse monoclonal antibody [11G7] to TLR2

Recommended Starting Dilutions: Flow Cytometry: 1/100 - 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: 11G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR