Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-THRB Mouse Monoclonal Antibody [clone: SPM222]

Supplier: Genetex
GTX17898
Thyroid hormone receptors (TRs) are ligand dependent members of the steroid/retinoic acid superfamily of transcription factors. The two genes encoding TRs identified to date, TRalpha and TRBeta, have been mapped to human chromosomes 17 and 3, respectively. TRs bind to thyroid hormone response elements (TREs) with half site binding motifs in the orientation of palindromes, direct repeats, or inverted palindromes. The affinities of binding are both variable and influenced differentially by 3,5,3?- triiodo-L-thyronine (T-3).

IHC-P: 1/50. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: THRB
Clonality: Monoclonal
Clone: SPM222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
250μl GTX17898 89351-424 Each (250µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol THRB
Antigen Name Thyroid Hormone Receptor beta
Reactivity Dog
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone SPM222
Host Mouse
Isotype IgG1
ImmunoChemistry Yes
Antigen Synonyms THRB2, NR1A2, GRTH, THR1, C-ERBA-2, THRB1, ERBA2, PRTH, C-ERBA-BETA
Amino Acid Number 31 to 50
Gene ID 7068
Concentration 0.2 mg/ml
Immunogen Synthetic peptide: GMSEACLHRKSHSERRSTLK, corresponding to N terminal amino acids 31-50 of Human Thyroid Hormone Receptor beta.
Purification Protein G purified
Sequence GMSEACLHRKSHSERRSTLK
Storage Buffer Preservative: Sodium Azide. Constituents: BSA, 10mM PBS. pH 7.4
Storage Temperature Store at 4ºC
Tested Applications IHC-P

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty