Anti-TG Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX73378
89353-170EA 357.89 USD
89353-170
Anti-TG Rabbit Polyclonal Antibody
Polyclonal Antibody to Human Thyroglobulin

This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Suitable for formalin/paraffin tissues.

Type: Primary
Antigen: TG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR