Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Thy1 Mouse Monoclonal Antibody (FITC) [clone: OX-7]

Supplier: Genetex

GTX20226
89360-804EA 358.53 USD InStock
89360-804
Anti-Thy1 Mouse Monoclonal Antibody (FITC) [clone: OX-7]
Antibodies
cell surface glycoprotein antigen [RGD]

Suggested working dilution for Flow cytometry: 1/10 - 1/100. Use 10 μl of the suggested working dilution to label 10^6 cells.

Type: Primary
Antigen: Thy1
Clonality: Monoclonal
Clone: OX-7
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty