Anti-TXNRD1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX13574
89359-758EA 581.69 USD
89359-758
Anti-TXNRD1 Goat Polyclonal Antibody
Antibodies
This protein is a pyridine nucleotide oxidoreductase which reduces thioredoxins as well as other substrates, and plays a role in selenium metabolism and protection against oxidative stress. It is thought to function as a homodimer which uses FAD as a cofactor. Each subunit contains a selenocysteine residue which is required for the catalytic activity. Alternative splicing results in several transcript variants encoding the same protein. Additional transcript variants have been described but their biological validity has not been determined.

Approx 60kDa band observed in Human Placenta lysates (predicted MW of 57kDa according to NP_003321). Recommended for use at 0.1-1µg/ml. This antibody is expected to recognise reported isoform 1 (NP_003321.2), isoform 2 (NP_877393.1; NP_877419.1; NP_877420.1) and isoform 3 (NP_001087240.1)

Type: Primary
Antigen: TXNRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR