Anti-THBS4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX87694
89366-942EA 616.5 USD
89366-942
Anti-THBS4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to THBS4

  • We recommend the following starting dilutions: IHC: 1:50~1:100 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: THBS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR