Anti-TGFB1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29758
89362-842EA 639.73 USD
89362-842
Anti-TGFB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TGF beta 1

assayed by ELISA against 0.1 ug of the immunizing peptide. A 1:1,000 to 1:3,000 dilution of the antibody is recommended for this assay.

Type: Primary
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR